Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

4-Methyl-2-thenoic acid 95%

Supplier: AMBEED, INC
Warning

A536224-10G A536224-1G A536224-25G A536224-5G
77116-774EA 60.77 USD InStock
77116-774 77116-776 77116-778 77116-780
4-Methyl-2-thenoic acid 95%
4-Methyl-2-thenoic acid
Precaución: for Research Use Only


Formula: C₆H₆O₂S
MDL Number: MFCD00089284
CAS Number: 14282-78-1
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty