Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

5-Benzyl-2-methyl-2,3,4,5-tetrahydro-1H-pyrido[4,3-b]indole heminaphthalene-1,5-disulfonate 98%

Supplier: AMBEED, INC
Warning

A545251-100MG A545251-1G A545251-1MG A545251-250MG A545251-25G A545251-5G
77116-844EA 9.27 USD InStock
77116-844 77116-846 77116-848 77116-850 77116-852 77116-854
5-Benzyl-2-methyl-2,3,4,5-tetrahydro-1H-pyrido[4,3-b]indole heminaphthalene-1,5-disulfonate 98%
5-Benzyl-2-methyl-2,3,4,5-tetrahydro-1H-pyrido[4,3-b]indole heminaphthalene-1,5-disulfonate
Precaución: for Research Use Only


Formula: C₄₈H₄₈N₄O₆S₂
MDL Number: MFCD00083420
CAS Number: 6153-33-9
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty