Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

8-Methyl-2,3,4,5-tetrahydro-1H-pyrido[4,3-b]indole hydrochloride 95+%

Supplier: AMBEED, INC
Warning

A565825-100MG A565825-1G A565825-250MG A565825-5G
77117-044EA 26.78 USD InStock
77117-044 77117-046 77117-048 77117-050
8-Methyl-2,3,4,5-tetrahydro-1H-pyrido[4,3-b]indole hydrochloride 95+%
8-Methyl-2,3,4,5-tetrahydro-1H-pyrido[4,3-b]indole hydrochloride
Precaución: for Research Use Only


Formula: C₁₂H₁₅ClN₂
MDL Number: MFCD07781100
CAS Number: 57933-28-5
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty