Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

LXR-623 98%

Supplier: AMBEED, INC
Warning

A566900-10MG A566900-1MG A566900-25MG A566900-50MG A566900-5MG
77117-062EA 114.33 USD InStock
77117-062 77117-064 77117-066 77117-068 77117-070
LXR-623 98%
LXR-623
Precaución: for Research Use Only


Formula: C₂₁H₁₂ClF₅N₂
MDL Number: MFCD16495812
CAS Number: 875787-07-8
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty