Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

(S)-2-(3-Aminopropanamido)-3-(1H-imidazol-5-yl)propanoic acid Zinc complex 97%

Supplier: AMBEED, INC
Warning

A580459-100G A580459-1G A580459-25G A580459-5G
77117-160EA 463.51 USD InStock
77117-160 77117-162 77117-164 77117-166
(S)-2-(3-Aminopropanamido)-3-(1H-imidazol-5-yl)propanoic acid Zinc complex 97%
(S)-2-(3-Aminopropanamido)-3-(1H-imidazol-5-yl)propanoic acid Zinc complex
Precaución: for Research Use Only


Formula: C₉H₁₁N₄O₃Zn
MW: 288.61 g/mol
MDL Number: MFCD00870906
CAS Number: 107667-60-7
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty