Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

4-[2-(Trifluoromethyl)phenoxy]piperidine hydrochloride 97%

Supplier: AMBEED, INC
Warning

A592921-10G A592921-1G A592921-5G
77117-224EA 140.07 USD InStock
77117-224 77117-226 77117-228
4-[2-(Trifluoromethyl)phenoxy]piperidine hydrochloride 97%
4-[2-(Trifluoromethyl)phenoxy]piperidine hydrochloride
Precaución: for Research Use Only


Formula: C₁₂H₁₅ClF₃NO
MDL Number: MFCD03840133
CAS Number: 823782-74-7
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty