Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

5-Acetylvaleric acid 98%

Supplier: AMBEED, INC
Danger

A597192-10G A597192-1G A597192-250MG A597192-25G A597192-5G
77117-240EA 62.83 USD InStock
77117-240 77117-242 77117-244 77117-246 77117-248
5-Acetylvaleric acid 98%
5-Acetylvaleric acid
Precaución: for Research Use Only


Formula: C₇H₁₂O₃
MW: 144.17 g/mol
MDL Number: MFCD00051479
CAS Number: 3128-07-2
UN: 3261
ADR: 8 Ⅱ
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty