Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

3-Chloropyridine 98%

Supplier: AMBEED, INC
Warning

A608351-1000G A608351-100G A608351-10G A608351-25G A608351-500G A608351-5G
77117-358EA 82.4 USD InStock
77117-358 77117-360 77117-362 77117-364 77117-366 77117-368
3-Chloropyridine 98%
3-Chloropyridine
Precaución: for Research Use Only


Formula: C₅H₄ClN
MDL Number: MFCD00006375
CAS Number: 626-60-8
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty