Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

2-Bromo-1-cyclopropylethanone 92%

Supplier: AMBEED, INC
Danger

A612728-1G A612728-5G A612728-10G
77117-386EA 22.66 USD InStock
77117-386 77594-880 77686-546
2-Bromo-1-cyclopropylethanone 92%
2-Bromo-1-cyclopropylethanone
Precaución: for Research Use Only


Formula: C₅H₇BrO
MDL Number: MFCD08460238
CAS Number: 69267-75-0
UN: 2810
ADR: 6.1,Ⅲ
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty