Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

1,3-Dibromo-2,5-dichlorobenzene 98%

Supplier: AMBEED, INC
Warning

A617890-100G A617890-10G A617890-25G A617890-5G
77117-414EA 110.21 USD InStock
77117-414 77117-416 77117-418 77117-420
1,3-Dibromo-2,5-dichlorobenzene 98%
1,3-Dibromo-2,5-dichlorobenzene
Precaución: for Research Use Only


Formula: C₆H₂Br₂Cl₂
MW: 304.7941 g/mol
MDL Number: MFCD00070784
CAS Number: 81067-41-6
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty