Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

3-Ethylaniline 98%

Supplier: AMBEED, INC
Danger

A624675-10G A624675-1G A624675-25G A624675-5G A624675-100G
77117-472EA 27.81 USD InStock
77117-472 77117-474 77117-476 77117-478 77686-566
3-Ethylaniline 98%
3-Ethylaniline
Precaución: for Research Use Only


Formula: C₈H₁₁N
MDL Number: MFCD00007818
CAS Number: 587-02-0
UN: 3082
ADR: 9,Ⅲ
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty