Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

4-Hydroxyphenylpyruvic acid 96%

Supplier: AMBEED, INC
Warning

A632541-1G A632541-250MG
77117-530EA 78.28 USD InStock
77117-530 77117-532
4-Hydroxyphenylpyruvic acid 96%
4-Hydroxyphenylpyruvic acid
Precaución: for Research Use Only


Formula: C₉H₈O₄
MDL Number: MFCD00002591
CAS Number: 156-39-8
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty