Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Agomelatine 98%

Supplier: AMBEED, INC
Warning

A643275-10MG A643275-1G A643275-25G A643275-25MG A643275-50MG A643275-5G A643275-5MG
77117-592EA 7.21 USD InStock
77117-592 77117-594 77117-596 77117-598 77117-600 77117-602 77117-604
Agomelatine 98%
Agomelatine
Precaución: for Research Use Only


Formula: C₁₅H₁₇NO₂
MW: 243.31 g/mol
MDL Number: MFCD00916659
CAS Number: 138112-76-2
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty