Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

2-(Hydroxymethyl)pyridin-4-ol 98%

Supplier: AMBEED, INC
Warning

A643773-100MG A643773-1G A643773-250MG
77117-606EA 15.45 USD InStock
77117-606 77117-608 77117-610
2-(Hydroxymethyl)pyridin-4-ol 98%
2-(Hydroxymethyl)pyridin-4-ol
Precaución: for Research Use Only


Formula: C₆H₇NO₂
MDL Number: MFCD13175434
CAS Number: 860411-74-1
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty