Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Ethyl 4-hydroxy-5,7-dimethylquinoline-3-carboxylate 95%

Supplier: AMBEED, INC
Warning

A651198-100MG A651198-1G A651198-250MG A651198-5G
77117-648EA 11.33 USD InStock
77117-648 77117-650 77117-652 77117-654
Ethyl 4-hydroxy-5,7-dimethylquinoline-3-carboxylate 95%
Ethyl 4-hydroxy-5,7-dimethylquinoline-3-carboxylate
Precaución: for Research Use Only


Formula: C₁₄H₁₅NO₃
MW: 245.274 g/mol
MDL Number: MFCD00173347
CAS Number: 93514-84-2
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty