Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Tioconazole 98%

Supplier: AMBEED, INC
Warning

A654539-1G A654539-1MG A654539-250MG A654539-25G A654539-5G
77117-674EA 24.73 USD InStock
77117-674 77117-676 77117-678 77117-680 77117-682
Tioconazole 98%
Tioconazole
Precaución: for Research Use Only


Formula: C₁₆H₁₃Cl₃N₂OS
MDL Number: MFCD00057276
CAS Number: 65899-73-2
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty