Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

1,2-Di-p-tolyldiazene 95%

Supplier: AMBEED, INC
Warning

A655858-10G A655858-1G A655858-250MG A655858-5G
77117-694EA 758.08 USD InStock
77117-694 77117-696 77117-698 77117-700
1,2-Di-p-tolyldiazene 95%
1,2-Di-p-tolyldiazene
Precaución: for Research Use Only


Formula: C₁₄H₁₄N₂
MW: 210.27 g/mol
MDL Number: MFCD00048089
CAS Number: 501-60-0
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty