Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

3,5-Dimethyl-4-ethoxyphenylboronic acid 98%

Supplier: AMBEED, INC
Warning

A663622-100G A663622-1G A663622-25G A663622-5G
77117-714EA 427.44 USD InStock
77117-714 77117-716 77117-718 77117-720
3,5-Dimethyl-4-ethoxyphenylboronic acid 98%
3,5-Dimethyl-4-ethoxyphenylboronic acid
Precaución: for Research Use Only


Formula: C₁₀H₁₅BO₃
MW: 194.04 g/mol
MDL Number: MFCD02179463
CAS Number: 850568-59-1
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty