Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

α,3-Dibromotoluene 98%

Supplier: AMBEED, INC
Danger

A672269-10G A672269-25G A672269-100G
77117-762EA 8.24 USD InStock
77117-762 77117-764 77686-636
α,3-Dibromotoluene 98%
α,3-Dibromotoluene
Precaución: for Research Use Only


Formula: C₇H₆Br₂
MDL Number: MFCD00000176
CAS Number: 823-78-9
UN: 3261
ADR: 8,Ⅱ
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty