Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

SB590885 98%

Supplier: AMBEED, INC
Warning

A720977-100MG A720977-10MG A720977-1MG A720977-50MG A720977-5MG
77118-038EA 761.17 USD InStock
77118-038 77118-040 77118-042 77118-044 77118-046
SB590885 98%
SB590885
Precaución: for Research Use Only


Formula: C₂₇H₂₇N₅O₂
MDL Number: MFCD16038645
CAS Number: 405554-55-4
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty