Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Mucobromic acid 98%

Supplier: AMBEED, INC
Danger

A723877-10G A723877-25G A723877-5G
77118-054EA 8.24 USD InStock
77118-054 77118-056 77118-058
Mucobromic acid 98%
Mucobromic acid
Precaución: for Research Use Only


Formula: C₄H₂Br₂O₃
MW: 257.88 g/mol
MDL Number: MFCD00063745
CAS Number: 488-11-9
UN: 3261
ADR: 8 Ⅲ
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty