Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

2-(Trifluoromethyl)pyrimidine 97%

Supplier: AMBEED, INC
Danger

A742431-100MG A742431-250MG
77118-130EA 42.23 USD InStock
77118-130 77118-132
2-(Trifluoromethyl)pyrimidine 97%
2-(Trifluoromethyl)pyrimidine
Precaución: for Research Use Only


Formula: C₅H₃F₃N₂
MW: 148.09 g/mol
MDL Number: MFCD10698646
CAS Number: 116470-67-8
UN: 1993
ADR: 3 Ⅲ
Order Now

 

ORDER

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty