Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

3-(3-(Benzyloxy)-4-methoxyphenyl)acrylic acid 95%

Supplier: AMBEED, INC
Warning

A796628-1G A796628-250MG A796628-25G A796628-5G
77118-352EA 22.65 USD InStock
77118-352 77118-354 77118-356 77118-358
3-(3-(Benzyloxy)-4-methoxyphenyl)acrylic acid 95%
3-(3-(Benzyloxy)-4-methoxyphenyl)acrylic acid
Precaución: for Research Use Only


Formula: C₁₇H₁₆O₄
MW: 284.30 g/mol
MDL Number: MFCD02684887
CAS Number: 24807-37-2
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty