Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

4,4'-Bis(ɑ,ɑ-dimethylbenzyl)diphenylamine 98%

Supplier: AMBEED, INC
Warning

A815973-100G A815973-25G A815973-500G A815973-5G
77118-402EA 13.39 USD InStock
77118-402 77118-404 77118-406 77118-408
4,4'-Bis(ɑ,ɑ-dimethylbenzyl)diphenylamine 98%
4,4'-Bis(alpha,alpha-dimethylbenzyl)diphenylamine
Precaución: for Research Use Only


Formula: C₃₀H₃₁N
MDL Number: MFCD00337918
CAS Number: 10081-67-1
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty