Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

(S)-1-Boc-2-isobutylpiperazine 97%

Supplier: AMBEED, INC
Warning

A830074-1G A830074-250MG
77118-428EA 48.41 USD InStock
77118-428 77118-430
(S)-1-Boc-2-isobutylpiperazine 97%
(S)-1-Boc-2-isobutylpiperazine
Precaución: for Research Use Only


Formula: C₁₃H₂₆N₂O₂
MDL Number: MFCD03787916
CAS Number: 674792-06-4
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty