Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

tert-Butyl (5-(hydroxymethyl)oxazol-2-yl)carbamate 95%

Supplier: AMBEED, INC
Warning

A839127-100MG A839127-1G A839127-250MG
77118-462EA 49.43 USD InStock
77118-462 77118-464 77118-466
tert-Butyl (5-(hydroxymethyl)oxazol-2-yl)carbamate 95%
tert-Butyl (5-(hydroxymethyl)oxazol-2-yl)carbamate
Precaución: for Research Use Only


Formula: C₉H₁₄N₂O₄
MW: 214.22 g/mol
MDL Number: MFCD12028213
CAS Number: 1142202-22-9
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty