Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Sodium-2-propylvalerate 97%

Supplier: AMBEED, INC
Warning

A841103-100G A841103-25G
77118-490EA 66.95 USD InStock
77118-490 77118-492
Sodium-2-propylvalerate 97%
Sodium-2-propylvalerate
Precaución: for Research Use Only


Formula: C₈H₁₅O₂Na
MW: 166.20 g/mol
MDL Number: MFCD00078604
CAS Number: 1069-66-5
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty