Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

1-(Benzyloxy)naphthalene 95%

Supplier: AMBEED, INC
Danger

A842573-100MG A842573-250MG
77118-500EA 29.87 USD InStock
77118-500 77118-502
1-(Benzyloxy)naphthalene 95%
1-(Benzyloxy)naphthalene
Precaución: for Research Use Only


Formula: C₁₇H₁₄O
MDL Number: MFCD00078325
CAS Number: 607-58-9
UN: 3077
ADR: 9,Ⅲ
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty