Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

α-Bromo-5-chloro-2-fluorotoluene 98%

Supplier: AMBEED, INC
Danger

A846276-10G A846276-25G A846276-5G
77118-524EA 18.54 USD InStock
77118-524 77118-526 77686-879
α-Bromo-5-chloro-2-fluorotoluene 98%
α-Bromo-5-chloro-2-fluorotoluene
Precaución: for Research Use Only


Formula: C₇H₅BrClF
MDL Number: MFCD01631437
CAS Number: 71916-91-1
UN: 3261
ADR: 8,Ⅱ
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty