Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

4-Phenoxyphenyl isocyanate 98%

Supplier: AMBEED, INC
Danger

A853414-1G A853414-250MG A853414-25G A853414-5G
77118-552EA 29.87 USD InStock
77118-552 77118-554 77118-556 77118-558
4-Phenoxyphenyl isocyanate 98%
4-Phenoxyphenyl isocyanate
Precaución: for Research Use Only


Formula: C₁₃H₉NO₂
MDL Number: MFCD00013876
CAS Number: 59377-19-4
UN: 2206
ADR: 6.1,Ⅲ
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty