Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

1-Bromo-2,5-difluoro-4-(trifluoromethyl)benzene 98%

Supplier: AMBEED, INC
Warning

A863441-1G A863441-250MG A863441-5G
77118-604EA 232.78 USD InStock
77118-604 77118-606 77118-608
1-Bromo-2,5-difluoro-4-(trifluoromethyl)benzene 98%
1-Bromo-2,5-difluoro-4-(trifluoromethyl)benzene
Precaución: for Research Use Only


Formula: C₇H₂BrF₅
MDL Number: MFCD01631615
CAS Number: 261945-75-9
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty