Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

DL-Isoserine 98%

Supplier: AMBEED, INC
Warning

A893693-10G A893693-25G A893693-5G A893693-100G A893693-1G
77118-752EA 27.81 USD InStock
77118-752 77118-754 77118-756 77686-934 77686-935
DL-Isoserine 98%
DL-Isoserine
Precaución: for Research Use Only


Formula: C₃H₇NO₃
MW: 105.09 g/mol
MDL Number: MFCD00008138
CAS Number: 565-71-9
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty