Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

N-(4-(2,4-Dimethylphenyl)thiazol-2-yl)benzamide 98%

Supplier: AMBEED, INC
Warning

A905052-10MG A905052-1MG A905052-50MG A905052-5MG
77118-784EA 17.52 USD InStock
77118-784 77118-786 77118-788 77118-790
N-(4-(2,4-Dimethylphenyl)thiazol-2-yl)benzamide 98%
N-(4-(2,4-Dimethylphenyl)thiazol-2-yl)benzamide
Precaución: for Research Use Only


Formula: C₁₈H₁₆N₂OS
MDL Number: MFCD01356769
CAS Number: 313553-47-8
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty