Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

2,4,6-Tris(4-fluorophenyl)pyrylium tetrafluoroborate 97%

Supplier: AMBEED, INC
Warning

A914155-100MG A914155-1G A914155-250MG
77118-846EA 72.1 USD InStock
77118-846 77118-848 77686-951
2,4,6-Tris(4-fluorophenyl)pyrylium tetrafluoroborate 97%
2,4,6-Tris(4-fluorophenyl)pyrylium tetrafluoroborate
Precaución: for Research Use Only


Formula: C₂₃H₁₄BF₇O
MDL Number: MFCD00160151
CAS Number: 62497-19-2
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty