Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

3-Aminocyclohexanecarboxylic acid (cis and trans mixture) 96%

Supplier: AMBEED, INC
Warning

A937168-1G A937168-250MG A937168-25G A937168-5G
77118-924EA 19.57 USD InStock
77118-924 77118-926 77118-928 77118-930
3-Aminocyclohexanecarboxylic acid (cis and trans mixture) 96%
3-Aminocyclohexanecarboxylic acid (cis and trans mixture)
Precaución: for Research Use Only


Formula: C₇H₁₃NO₂
MDL Number: MFCD00059562
CAS Number: 25912-50-9
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty