Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

(R)-4-Cyanothiazolidine hydrochloride 98%

Supplier: AMBEED, INC
Warning

A943446-100MG A943446-1G A943446-250MG
77118-950EA 75.19 USD InStock
77118-950 77118-952 77118-954
(R)-4-Cyanothiazolidine hydrochloride 98%
(R)-4-Cyanothiazolidine hydrochloride
Precaución: for Research Use Only


Formula: C₄H₆N₂S·ClH
MDL Number: MFCD02683061
CAS Number: 391248-17-2
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty