Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

(S)-1-(3-Pyridyl)ethanol 98%

Supplier: AMBEED, INC
Warning

A958941-100MG A958941-1G A958941-250MG A958941-5G
77118-996EA 31.93 USD InStock
77118-996 77118-998 77119-000 77119-002
(S)-1-(3-Pyridyl)ethanol 98%
(S)-1-(3-Pyridyl)ethanol
Precaución: for Research Use Only


Formula: C₇H₉NO
MDL Number: MFCD07368884
CAS Number: 5096-11-7
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty