Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

α-Bromo-4-chloro-2-fluorotoluene 98%

Supplier: AMBEED, INC
Danger

A964216-1G A964216-25G A964216-5G
77119-026EA 11.33 USD InStock
77119-026 77119-028 77119-030
α-Bromo-4-chloro-2-fluorotoluene 98%
α-Bromo-4-chloro-2-fluorotoluene
Precaución: for Research Use Only


Formula: C₇H₅BrClF
MW: 223.47
MDL Number: MFCD00831105
CAS Number: 71916-82-0
UN: 1760
ADR: 8 Ⅱ
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty