Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

(S)-2-Methylpyrrolidine 96%

Supplier: AMBEED, INC
Danger

A973309-10G A973309-1G A973309-25G A973309-5G
77119-062EA 264.71 USD InStock
77119-062 77119-064 77119-066 77119-068 77119-070
(S)-2-Methylpyrrolidine 96%
(S)-2-Methylpyrrolidine
Precaución: for Research Use Only


Formula: C₅H₁₁N
MDL Number: MFCD07779425
CAS Number: 59335-84-1
UN: 2924
ADR: 3,8,Ⅱ
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty