Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

N-Isopropylbenzylamine 98%

Supplier: AMBEED, INC
Warning

A996247-100G A996247-25G A996247-500G
77119-124EA 35.02 USD InStock
77119-124 77119-126 77119-128
N-Isopropylbenzylamine 98%
N-Isopropylbenzylamine
Precaución: for Research Use Only


Formula: C₆H₅CH₂N(CH₃)(CH₂CH₃)
MDL Number: MFCD00008863
CAS Number: 102-97-6
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty