Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

2,4-Difluoro-3-methylbenzonitrile 95%

Supplier: AMBEED, INC
Danger

A100609-10G A100609-1G A100609-250MG A100609-5G
77135-446EA 117.42 USD InStock
77135-446 77135-448 77135-450 77135-452
2,4-Difluoro-3-methylbenzonitrile 95%
2,4-Difluoro-3-methylbenzonitrile
Precaución: for Research Use Only


Formula: C₈H₅F₂N
MDL Number: MFCD07777155
CAS Number: 847502-87-8
UN: 3439
ADR: 6.1,Ⅲ
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty