Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Methyl-o-toluate 98%

Supplier: AMBEED, INC
Warning

A102579-100G A102579-25G A102579-5G
77135-558EA 12.36 USD InStock
77135-558 77135-560 77135-562 77135-564
Methyl-o-toluate 98%
Methyl-o-toluate
Precaución: for Research Use Only


Formula: C₉H₁₀O₂
MW: 150.18
Boiling Pt: 215
Melting Pt: -50
Density: 1.079
MDL Number: MFCD00008428
CAS Number: 89-71-4
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty