Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

(R)-2-Aminopropanamide 95%

Supplier: AMBEED, INC
Warning

A105869-10G A105869-1G A105869-250MG A105869-25G A105869-5G
77135-696EA 157.59 USD InStock
77135-696 77135-698 77135-700 77135-702 77135-704
(R)-2-Aminopropanamide 95%
(R)-2-Aminopropanamide
Precaución: for Research Use Only


Formula: C₃H₈N₂O
MDL Number: MFCD00672501
CAS Number: 35320-22-0
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty