Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

3-Bromo-5-(1,1-difluoroethyl)pyridine 95%

Supplier: AMBEED, INC
Warning

A108735-100MG A108735-1G A108735-250MG
77135-842EA 38.12 USD InStock
77135-842 77135-844 77135-846
3-Bromo-5-(1,1-difluoroethyl)pyridine 95%
3-Bromo-5-(1,1-difluoroethyl)pyridine
Precaución: for Research Use Only


Formula: C₇H₆BrF₂N
MDL Number: MFCD18254770
CAS Number: 1108724-32-8
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty