Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Cyclohexen-1-ylbenzene 97%

Supplier: AMBEED, INC
Warning

A108785-100G A108785-10G A108785-1G A108785-25G A108785-5G
77135-848EA 246.17 USD InStock
77135-848 77135-850 77135-852 77135-854 77135-856
Cyclohexen-1-ylbenzene 97%
Cyclohexen-1-ylbenzene
Precaución: for Research Use Only


Formula: C₁₂H₁₄
MDL Number: MFCD00001542
CAS Number: 771-98-2
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty