Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

4-Chloro-2-methoxy-6-nitroaniline 97%

Supplier: AMBEED, INC
Warning

A110486-100MG A110486-10G A110486-1G A110486-250MG A110486-5G
77136-002EA 6.18 USD InStock
77136-002 77136-004 77136-006 77136-008 77136-010
4-Chloro-2-methoxy-6-nitroaniline 97%
4-Chloro-2-methoxy-6-nitroaniline
Precaución: for Research Use Only


Formula: C₇H₇ClN₂O₃
MDL Number: MFCD28405560
CAS Number: 859877-49-9
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty