Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

2-Bromo-4-(trifluoromethoxy)aniline 98%

Supplier: AMBEED, INC
Warning

A110990-100G A110990-10G A110990-25G A110990-5G
77136-030EA 54.59 USD InStock
77136-030 77136-032 77136-034 77136-036
2-Bromo-4-(trifluoromethoxy)aniline 98%
2-Bromo-4-(trifluoromethoxy)aniline
Precaución: for Research Use Only


Formula: C₇H₅BrF₃NO
MW: 256.02
MDL Number: MFCD00203478
CAS Number: 175278-17-8
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty