Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

(R)-1,2-Butanediol 98%

Supplier: AMBEED, INC
Warning

A114219-10G A114219-1G A114219-25G A114219-5G
77136-172EA 56.65 USD InStock
77136-172 77136-174 77136-176 77136-178
(R)-1,2-Butanediol 98%
(R)-1,2-Butanediol
Precaución: for Research Use Only


Formula: C₄H₁₀O₂
MDL Number: MFCD09953765
CAS Number: 40348-66-1
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty