Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Diethyl isophthalate 98%

Supplier: AMBEED, INC
Warning

A116329-100G A116329-10G A116329-25G A116329-5G
77136-284EA 41.19 USD InStock
77136-284 77136-286 77136-288 77136-290
Diethyl isophthalate 98%
Diethyl isophthalate
Precaución: for Research Use Only


Formula: C₁₂H₁₄O₄
MW: 222.23 g/mol
MDL Number: MFCD01717556
CAS Number: 636-53-3
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty