Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

(R)-(-)-1-Indanol 95%

Supplier: AMBEED, INC
Warning

A116744-100MG A116744-1G A116744-250MG A116744-5G
77136-310EA 31.94 USD InStock
77136-310 77136-312 77136-314 77136-316
(R)-(-)-1-Indanol 95%
(R)-(-)-1-Indanol
Precaución: for Research Use Only


Formula: C₉H₁₀O
MDL Number: MFCD00064164
CAS Number: 697-64-3
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty